Eisco® Joint Models | Avantor
Eisco® Joint Models
Supplier:   Wards
Eisco® Joint Models
Supplier:  Wards
Certificates
Eisco® Joint Models
Eisco® Joint Models
4 Options Available Below
Product Details & Documents
Study How Joints Are Held Together.
  • Shows ligaments of 4 common joints
  • Ligaments are slightly flexible, and joints are reinforced to minimize damage
  • Mounted on base
These joint models from Eisco are a great addition to your anatomy lessons. Joint structure is easily visible with both bone and ligaments visible. The ligament material has some resilience, and the joints are reinforced to reduce the chance of damage to the model while using. This limits the range of motion for each joint, but still allows your students to see how movement is transferred through the ligaments.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Hip Joint
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $193.15
    $193.15
    Each
       $193.15
    La quantité minimale de commande est de 1
    Supplier Number: AM0135
  • Elbow Joint
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $193.15
    $193.15
    Each
       $193.15
    La quantité minimale de commande est de 1
    Supplier Number: AM0136
  • Shoulder Joint
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $207.60
    $207.60
    Each
       $207.60
    Supplier Number: AM133AS
  • Knee Joint
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $164.00
    $164.00
    Each
       $164.00
    Supplier Number: AM0134AS