SP Bel-Art Screw Cap Jars, Polypropylene, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
White polypropylene jars and screw caps in an assortment of sizes are FDA approved and are excellent for storage of liquid or dry stock, or for use with samples.
- White polypropylene construction
- Suited for liquid or dry stock storage
- Autoclavable
Jars are autoclavable at 121˚C (250˚F). Screw caps, with PE foam lining, are not autoclavable.
Frequently Bought Together