VWR® Illuminated Tissue Flotation Baths
VWR® Illuminated Tissue Flotation Baths
VWR® Illuminated Tissue Flotation Baths
VWR® Illuminated Tissue Flotation Baths
The VWR® Illuminated tissue flotation baths are designed for use in the Histology Lab that is conducting tissue preparation.
  • LED illuminated black background
  • Easy removal of sample for cleaning
  • Compact footprint
  • Easy-to-read display
An LED illuminated black background with a transparent PYREX® dish allows for easy viewing of the sample which can also be removed for easy cleaning
A compact footprint uses less bench space and the easy-to-read display can be seen easily through out the lab.
The digital microprocessor control and temperature sensor ensures temperature uniformity while the safety over-temp device provides protection for of the samples. VWR® offers both a Premium version and a Standard version of these Tissue Flotation Baths. The Premium version includes both an Orienter Block and a Dryer Block on the back of the unit which assists in the drying and smoothing of slides and specimens.
 : UL/CSA Listed and CE Certified.
 : The VWR® Illuminated Tissue Flotation Bath is supplied with one PYREX® Dish, Instruction Manual and Power Cord. Each unit comes with a two-year warranty on parts and labor. Additional PYREX® dishes are available for purchase.
  • Catalog No:
  • 97043-530
  • 97043-534
  • Capacity:
  • 1.9 L (2 qt.)
  • 1.9 L (2 qt.)
  • Temperature range:
  • Ambient +5°C to 65°C
  • Ambient +5°C to 65°C
  • Temperature uniformity:
  • +/- 1°C at 55°C
  • +/- 1°C at 55°C
  • Block temperature:
  • Dryer preset at 40 °C +/- 3 °C; Orienter fixed at 55 °C +/- 3 °C
  • Electrical:
  • 115V, 50/60Hz, 272W
  • 115V, 50/60Hz, 272W
  • Ext. W×D×H:
  • 14.25×14.25×4.5"
  • 14.25×14.25×4.5"
  • Int. W×D×H:
  • 7×11×1.7" (dish)
  • 7×11×1.7" (dish)
  • Shipping weight:
  • 12 lbs.
  • 12 lbs.
Product Family Options
  • Désignation
  • Illuminated Tissue Flotation Bath, 115V, 50/60Hz
    Each
    Each - $5,660.57
    $5,660.57
    Each
       $5,660.57
    Product available on GSA Advantage®
    Specifications
    Cat. No.
    97043-530
    Capacity
    1.9 L (2 qt.)
    Temperature range
    Ambient +5°C to 65°C
    Temperature uniformity
    +/- 1°C at 55°C
    Block temperature
    Electrical
    115V, 50/60Hz, 272W
    Ext. W×D×H
    14.25×14.25×4.5"
    Int. W×D×H
    7×11×1.7" (dish)
    Shipping weight
    12 lbs.
    Product available on GSA Advantage®
  • Premium illuminated tissue flotation bath, 115V, 50/60Hz
    Each
    Each - $4,492.63
    $4,492.63
    Each
       $4,492.63
    Product available on GSA Advantage®
    Specifications
    Cat. No.
    97043-534
    Capacity
    1.9 L (2 qt.)
    Temperature range
    Ambient +5°C to 65°C
    Temperature uniformity
    +/- 1°C at 55°C
    Block temperature
    Dryer preset at 40 °C +/- 3 °C; Orienter fixed at 55 °C +/- 3 °C
    Electrical
    115V, 50/60Hz, 272W
    Ext. W×D×H
    14.25×14.25×4.5"
    Int. W×D×H
    7×11×1.7" (dish)
    Shipping weight
    12 lbs.
    Product available on GSA Advantage®
Accessories
  •  
    Description
  • 13278-282
    Dish for use with Illuminated Tissue Flotation Baths
    Each - $197.81