Model AL-AP Alkalinity Test Kit, Hach
Fournisseur: Hach
Certificats
À propos de cet article
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists
This kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
Utilizing drop count titration/sulfuric acid to determine alkalinity, this kit contains 100 tests and is a simple, economical titration method.
Ordering information: Kit includes sample measuring tube, mixing bottle, reagents, instructions and carrying case.
Frequently Bought Together