Pool Master Test Kit, Hach | Avantor
Pool Master Test Kit, Hach
Supplier:   Hach
Pool Master Test Kit, Hach
Supplier:  Hach
Certificates
Pool Master Test Kit, Hach
Pool Master Test Kit, Hach
1 Options Available Below
Product Details & Documents
Hach's Pool Master Test Kit combines several tests into a single test kit. The test kit includes tests for alkalinity, chlorine, hardness, and pH, plus specialized tests for total alkalinity, calcium hardness, and cyanuric acid.
Uses simple, economical drop count titration and color disc methods.
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Ordering information: Ten reagents, clippers, graduated cylinder, sample cell with cap, dipstick, two color discs, viewing tubes, comparator box, instructions and carrying case included.
Product Family Options
Informations sur le produit
  • Désignation
    Range
    Availability
    Price
  • Pool Master Test Kit
    6.6 - 8.4 pH
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $663.84
    $663.84
    Each
       $663.84
    Supplier Number: 2315001
  • Pool Master Test Kit
    6.6 - 8.4 pH
    Discontinued
    Supplier Number: -