Pool Master Test Kit, Hach
À propos de cet article
Hach's Pool Master Test Kit combines several tests into a single test kit. The test kit includes tests for alkalinity, chlorine, hardness, and pH, plus specialized tests for total alkalinity, calcium hardness, and cyanuric acid.
Uses simple, economical drop count titration and color disc methods.
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Ordering information: Ten reagents, clippers, graduated cylinder, sample cell with cap, dipstick, two color discs, viewing tubes, comparator box, instructions and carrying case included.
1 Options Available Below
Product Details & Documents
Hach's Pool Master Test Kit combines several tests into a single test kit. The test kit includes tests for alkalinity, chlorine, hardness, and pH, plus specialized tests for total alkalinity, calcium hardness, and cyanuric acid.
Uses simple, economical drop count titration and color disc methods.
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Ordering information: Ten reagents, clippers, graduated cylinder, sample cell with cap, dipstick, two color discs, viewing tubes, comparator box, instructions and carrying case included.
Product Family Options
Informations sur le produit
- DésignationRangeAvailabilityPrice
- Supplier Number: 2315001
- Pool Master Test Kit6.6 - 8.4 pHDiscontinued-Supplier Number: -