VWR® Traceable® Extra-Extra Large Digit Timer | Avantor
VWR® Traceable® Extra-Extra Large Digit Timer
Extra-extra large digit timer, 2¾×3⅛×1"
Catalog # 89369-164
Supplier:   VWR International
VWR® Traceable® Extra-Extra Large Digit Timer
Catalog # 89369-164
Supplier:  VWR International
VWR® Traceable® Extra-Extra Large Digit Timer
VWR® Traceable® Extra-Extra Large Digit Timer
Extra-extra large digit timer, 2¾×3⅛×1"
Price
$82.84
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Extra large 1¹/₂" digit size allows for easy viewing from up to 60 feet away.
  • Easy to use
  • Automatic timing memory
  • Large, easy-to-read display
Reliable, 3-key operation makes countdown settings in 1 second increments easy to program. Timer countdowns and alarms on single channel for ease-of-use. Counts up during alarm function indicating elapsed time since alarming. Times up to 99 minutes, 59 seconds with one second resolution. Automatic Bounceback(TM) Memory for repetitive timing. Accuracy is 0.01%. Counts up in stopwatch mode.
Certifications: To assure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Ordering information: Supplied with wall mount clip, bench stand, magnetic back, battery and Traceable® Certificate.
Spécifications
  • Désignation:
    Extra-extra large digit timer
  • W×H×D:
    2¾×3⅛×1"