CL17 DPD Indicator, Hach
CL17 DPD Indicator, Hach
24 g, DPD (N,N-diethyl-p-phenylenediamine), Jar, 450
Catalog # CA22972-55
Supplier:   Hach
Supplier #:  2297255
CL17 DPD Indicator, Hach
Catalog # CA22972-55
Supplier:  Hach
CAS Number:  
CL17 DPD Indicator, Hach
CL17 DPD Indicator, Hach
24 g, DPD (N,N-diethyl-p-phenylenediamine), Jar, 450
CAS Number:  
Price
$80.19
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
DPD is an indicator that is used to measure either Free Chlorine or Total Chlorine with the CL17 Analyzer. Depending upon whether Free Chlorine or Total Chlorine is being measured, the DPD indicator is added to Free Chlorine or Total Chlorine Indicator Solution and used with Free or Total Buffer Solution by the CL17 Analyzer. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
  • Quality, Reliability
  • Simple, Predictable Maintenance
  • Reagent Delivery Program
Spécifications
Spécifications
  • Méthode chimique:
    DPD (N,N-diethyl-p-phenylenediamine)
  • Type de conditionnement:
    Jar
  • Taille:
    24 g
  • No. of Tests:
    450