Screw Caps with Partial Slit Septa for 12×32 mm Vials, Microsolv Technology Corp
Catalog # 97051-404
Supplier: MicroSolv Technology Corp
Specifications
- Color:Blue
- Description:Polypropylene Screw Cap with Silicone/PTFE Septa for 12 x 32 mm Vials, 9-425 Thread
- Cat. no.:97051-404
- Supplier No.:9502S-34C-B
Specifications
About this item
Screw Caps have a partial precut Y shape slit to prevent evaporation of volatile samples before injection
Helps to minimize vacuum forming in the vial.