ART™ Barrier Reload Insert Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
The ART™ (Aerosol Resistant Tip) Barrier Reload Insert pipette tips seal when exposed to potential contaminants, providing accurate and consistent performance.
The ART inserts are environmentally friendly. Individually packed blister tubs are made out of recycled plastic and are designed to be used with the reusable ART hinged racks - creating the ART Reload System. The pipette tips from the reload inserts clip securely into the hinged racks for efficient reloading.
Specifications
- Catalog No:
- 89082-130
- 89082-146
- 89082-144
- 89082-142
- 89082-140
- 89082-138
- 89082-136
- 89082-134
- 89082-132
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)