Stablcal® Turbidity Standards Calibration Kit, 2100P Portable Turbidimeter, 500 ml bottles, Hach
Catalogue # CA97026-814
Fournisseur: Hach
Numéro CAS:
Spécifications
- Turbidity:100 NTU800 NTU< 0.1 NTU20 NTU
- Size:4×500 ml bottle
- Cat. no.:CA97026-814
- Supplier No.:2659400
Spécifications
À propos de cet article
Stablcal Calibration Standards Kit, for 2100P Portable Turbidimeter. USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. Kit includes 500 ml bottles of <0.1, 20, 100 and 800 NTU standard.
- Precise, Pre-mixed Formazin Solutions
- Stablcal is Safe to Handle
- No special glassware or difficult lab techniques
- Developed for use in any turbidimeter
- Enhanced Stability Saves Time and Labor