Sample Cell Oiling Cloth, Hach | Avantor
Sample Cell Oiling Cloth, Hach
Catalog # CA47076-00
Supplier:   Hach
Fournisseur #:  4707600
Sample Cell Oiling Cloth, Hach
Catalog # CA47076-00
Supplier:  Hach
Sample Cell Oiling Cloth, Hach
Sample Cell Oiling Cloth, Hach
Price
$79.68
Log in pour la disponibilité
Each
Product Details & Documents
Use to apply silicone oil to turbidimeter sample cells.