BOD Nutrient Buffer Pillows, Hach | Avantor
BOD Nutrient Buffer Pillows, Hach
Supplier:   Hach
BOD Nutrient Buffer Pillows, Hach
Supplier:  Hach
Certificates
BOD Nutrient Buffer Pillows, Hach
BOD Nutrient Buffer Pillows, Hach
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For preparation of BOD5 dilution water. SP based.
Each pillow prepares 3 or 19 liters of dilution water.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Volume
    Final Volume
    Availability
    Price
  • BOD Nutrient Buffer Pillows
    50 pillows
    3 ml
    3 L
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50 (3ml)
    Pack of 50 (3ml) - $144.20
    $144.20
    Pack of 50 (3ml)
       $144.20
    Supplier Number: 1486166
  • BOD Nutrient Buffer Pillows
    25 pillows
    19 ml
    19 L
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25 (19ml)
    Pack of 25 (19ml) - $221.95
    $221.95
    Pack of 25 (19ml)
       $221.95
    Supplier Number: 1486398