SP Bel-Art Poxygrid® 'Rack And A Half Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
High-quality epoxy-coated wire racks in convenient extra-long sizes for volume test tube work.
- Resistant to chemicals and corrosion
- Autoclavable
- Epoxy-coated steel wire construction
- Conveniently sized
Epoxy-coated steel wire is chemical and corrosion resistant and steam autoclavable at 121°C (250°F).
Frequently Bought Together