Nunc® Internally-Threaded Universal Tubes and Rack System, 2.0 ml, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Nunc® 2.0 ml internally-threaded universal tubes are available racked or in bulk, with non-coded or 2D barcodes to serve every need.
- All tubes feature a white marking area and graduations
Part of the universal system, this tube offers the best features of the Thermo Scientific storage tube portfolio: universal cap, universal latch rack, and free-standing tube design. The tubes and rack are ideal for both lower-throughput benchtop use and high-throughput automation use.
Certifications: IATA certified.
Frequently Bought Together