VWR® Bin Liners
VWR® Bin Liners
   Options durables disponibles
Supplier:   VWR International
VWR® Bin Liners
Supplier:  VWR International
   Options durables disponibles
Certificates
VWR® Bin Liners
5 Options Available Below
Product Details & Documents
LDPE liners for use in clean and controlled aseptic environments.
  • Available non sterile or gamma irradiated
Uses include transport of product and waste disposal.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 10753-432
  • 75840-126
  • 89495-442
  • 78107-336
  • 78107-338
  • Environmentally Preferable:
  • Waste Reducing
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Matière
    Dimensions
    Épaisseur
    Packaging
    Availability
    Price
  • Bin liner
    Clear
    LDPE
    24×36"
    3 mil
    Double-bagged in groups of 5
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $331.52
    $331.52
    Case of 50
       $331.52
    Spécifications:
    Small Business Enterprise
      Waste Reducing
    More
    Specifications
    Cat. No.
    10753-432
    Environmentally Preferable
    Waste Reducing
    Small Business Enterprise
      Waste Reducing
  • Gamma irradiated bin liner
    Clear
    LDPE
    34×40"
    4 mil
    Individually bagged and tagged in a double bagged pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $481.90
    $481.90
    Case of 50
       $481.90
    Spécifications:
    More
    Specifications
    Cat. No.
    75840-126
    Environmentally Preferable
  • Gamma irradiated bin liner
    Clear
    LDPE
    40×48"
    2 mil
    Individually bagged and tagged in a double bagged pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $613.04
    $613.04
    Log in pour la disponibilité
    Pallet of 2000
    Pallet of 2000 - $24,521.60
    $24,521.60
    Case of 50
       $613.04
    Pallet of 2000
       $24,521.60
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89495-442
    Environmentally Preferable
    Small Business Enterprise
  • BioHazard bin liner, flat bottom
    Red
    LDPE
    38×48"
    3 mil
    Double-bagged in groups of 5
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $267.21
    $267.21
    Case of 50
       $267.21
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78107-336
    Environmentally Preferable
    New Product
  • BioHazard bin liner, flat bottom, irradiated
    Red
    LDPE
    38×48"
    3 mil
    Individually bagged and tagged in a double bagged pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $646.58
    $646.58
    Case of 50
       $646.58
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78107-338
    Environmentally Preferable