VWR® Microfiber String Mop
VWR® Microfiber String Mop
VWR® Microfiber String Mop
VWR® Microfiber String Mop
This Microfiber String Mop is an absorbent and hard wearing mop for cleaning and liquid pick-up in controlled and critical environments.
  • Highly absorbent
  • Durable
The unique ribbing of the microfiber strands produces a mop that is both low linting and absorbent. The strands are comprised of split fiber polyester filaments, 30 times smaller than cotton fibers. These microfibers work to pick up tiny particles where cotton/rayon blends and polyesters may often roll over the dirt. The solid constructions of the mop, combined with the durable microfiber, allow the mop to be used with strong disinfectants, laundered and re-used.
Product Family Options
  • Désignation
    Weight
  • Microfiber string mop
    12 oz.
  • Microfiber string mop
    16 oz.
    Each
    Each - $92.67
    $92.67
    Case of 24
    Case of 24 - $2,224.08
    $2,224.08
    Each
       $92.67
    Case of 24
       $2,224.08
  • Microfiber string mop, irradiated
    16 oz.
    Case of 24
    Case of 24 - $1,657.31
    $1,657.31
    Case of 24
       $1,657.31