ContactUsLinkComponent
GreenHouse Blowdown Evaporator, System 18, Heidolph Radleys
GreenHouse Blowdown Evaporator, System 18, Heidolph Radleys
Catalog # CA11008-876
Supplier:  Heidolph NA, LLC
GreenHouse Blowdown Evaporator, System 18, Heidolph Radleys
Catalog # CA11008-876
Supplier:  Heidolph NA, LLC
Supplier Number:  015850180CA
Specifications
  • Ø:
    16 mm well
  • Temperature Range:
    Heating ambient to 150 °C
  • Description:
    System 18 with 24 reaction positions (MTP format), 30 minute max. heat-up time
  • Cat. no.:
    CA11008-876
  • Working Volume:
    0.5 - 7.0 ml
  • Speed Range:
    0 - 1100 rpm
  • Temperature Control Accuracy:
    Better than 0.5 °C
  • Supplier No.:
    015850180CA
Specifications
About this item
Turn your existing GreenHouse Parallel Synthesizer into a parallel evaporator, or simply use this system as your own personal benchtop evaporator
The GreenHouse Blowdown uses a precisely controlled flow of inert gas combined with digitally controlled heating to carefully evaporate your samples. Interchangeable plates with either 8 or 24 hollow pins deliver an equal flow of gas to each tube, vial or well. The absence of any vacuum avoids bumping, further protecting your sample during evaporation.
Ordering information: This System includes GreenHouse Blowdown head, Long Blowdown pin plate 6 x 4 for GreenHouse tubes, Blowdown condenser, Blowdown 150 mL reservoir, Blowdown pin plat 6 x 4, Blowdown stand-alone base, GreenHouse glass cylinder, AT02 vial rack 6 x 4 for 13 mm OD vials. Inert gas: Yes; Reflux: Yes.