Hamilton Syringes for CTC Analytics Sampler Handlers, MicroLiter
À propos de cet article
The design of the C-line is based on the improvements employed in CTC’s X-Type syringe
Excluded are the inert needle material and the specially treated inside of the glass barrel. As with the X-Type syringe the construction is characterized by a direct glass/needle connection that completely avoids sample contact with glue or cement. A long-life polymer compound of the latest generation is used as plunger tip material. Additionally, a new flange design with adjustable plunger protects the tip from squeezing.
22 Options Available Below
Product Details & Documents
The design of the C-line is based on the improvements employed in CTC’s X-Type syringe
Excluded are the inert needle material and the specially treated inside of the glass barrel. As with the X-Type syringe the construction is characterized by a direct glass/needle connection that completely avoids sample contact with glue or cement. A long-life polymer compound of the latest generation is used as plunger tip material. Additionally, a new flange design with adjustable plunger protects the tip from squeezing.
Product Family Options
Informations sur le produit
- DésignationVolumeJaugePoint StyleAvailabilityPrice
- Supplier Number: XT-G10022S-3
- XT-Syringe25 µl22s3Discontinued-Supplier Number: -
- Supplier Number: XT-G5022S-3
- Supplier Number: XT-G10022-3
- Supplier Number: CL-L1.226P-AS
- Supplier Number: CL-L526S-AS
- Supplier Number: CL-L1026S-2
- Supplier Number: CL-L1026S-AS
- Supplier Number: CL-G1022S-3
- Supplier Number: CL-G2522S-3
- Supplier Number: CL-G2526S-AS
- Supplier Number: CL-G10022S-3
- Supplier Number: CL-G10026S-AS
- Supplier Number: CL-G25022-3
- Supplier Number: CL-G25026-AS
- C-Line Syringe500 µl223Discontinued-Supplier Number: -
- Supplier Number: CL-G50026-AS
- Supplier Number: CL-G100022-3
- Supplier Number: CL-G250022-3
- Supplier Number: HS-1.0235
- Supplier Number: HS-2.5235
- Supplier Number: HS-1.0265
- Supplier Number: HS-2.5265
- Supplier Number: HS-5.0235
Plungers
Informations sur le produit
- Catalog #DésignationAvailabilityPrice