Microcentrifuge Tubes, Graduated, with Secure-Lock™ MCT Seals, Simport Scientific
Supplier: Simport Scientific
Certificates
About this item
Secure-Lock™ polypropylene microcentrifuge tubes offer a special locking clasp to produce a more secure closure.
- Autoclavable to 121 °C
- Pierceable lid
- Etched surface on side of tube for sample identification
This will help to prevent tubes from opening during centrifugation, boiling, storing, freezing and shipping. They are graduated at 0.5, 1.0 and 1.5 ml.
Certifications: Certified RNase-, DNase-, Pyrogen- and DNA-free.
Ordering information: Packaged in tamperproof resealable safety-lock bags.
Frequently Bought Together