SlideRite™ Water Soluble Spray
CA11027-246
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:SlideRite™ Water Soluble Spray
- Cat. no.:CA11027-246
- Supplier No.:1801
Slide Rite is a highly concentrated, water-soluble spray that forms a lasting protective coating over all types of cytological smears
Highly concentrated, water-soluble spray
- For use on cytological smears
- 1.5 oz. spray bottle
Many cytological specimens are fragile and degenerate quickly. Therefore an immediate fixation of the material to be studied is essential.