ContactUsLinkComponent
LeStab® Inocula Needles, Decon Labs
LeStab® Inocula Needles, Decon Labs
Catalog # CA11027-278
Supplier:  Decon Labs
undefined
LeStab® Inocula Needles, Decon Labs
Catalog # CA11027-278
Supplier:  Decon Labs
Supplier Number:  MP1865
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Inocula needle
  • Cat. no.:
    CA11027-278
  • Handle Color:
    Clear
  • Supplier No.:
    MP1865
Specifications
About this item
LeStab® Inocula needles are used to inoculate or transfer microbiological samples.
  • Nichrome wire cools rapidly after flaming
  • Decon also offers 3" inoculating needles
  • Available in convenient packaging configurations
  • Custom packaging configurations may be available
Decon LeStab® inoculating needles are made of high-quality nichrome wire, which is designed to cool rapidly after flaming. Most needles are permanently attached to lightweight, insulated handles which are perfectly balanced for easy manipulation.
These needles fit in Decon’s LeLoop® brass holders. Brass holders come in 6 and 8" sizes.