The Pill™ Autoclave Deoderizers
The Pill™ Autoclave deodorizer capsules contain concentrated liquid odor neutralizers which absorb odors instead of just masking them.
Gelatin capsule keeps deodorizing liquid off hands and clothes
- Capsules facilitate accurate unit dosing
- Pleasant fresh scent
One capsule deodorizes 7 cubic feet of autoclave capacity with full load of soiled labware. Gelatin capsules are not suited to small tabletop autoclaves. One capsule is too strong for small tabletop autoclaves. A pungent residual odor of unused deodorizer will be present in units smaller than 5 cubic feet.
The Pill™ Autoclave deodorizer capsules contain concentrated liquid odor neutralizers which absorb odors instead of just masking them.
Gelatin capsule keeps deodorizing liquid off hands and clothes
- Capsules facilitate accurate unit dosing
- Pleasant fresh scent
One capsule deodorizes 7 cubic feet of autoclave capacity with full load of soiled labware. Gelatin capsules are not suited to small tabletop autoclaves. One capsule is too strong for small tabletop autoclaves. A pungent residual odor of unused deodorizer will be present in units smaller than 5 cubic feet.
Product Family Options
- Désignation
- Autoclave Deoderizer-: 5100
- Autoclave DeoderizerPack of 3,000$515.00Case of 18000$3,090.00: 5500