Certipur® Anion multi-element standard I | Avantor
Certipur® Anion multi-element standard I
500 ml, Plastic bottle
Catalog # CA1.11437.0500
Supplier:   MilliporeSigma
Fournisseur #:  1.11437.0500
Certipur® Anion multi-element standard I
Catalog # CA1.11437.0500
Supplier:  MilliporeSigma
CAS Number:  
Certipur® Anion multi-element standard I
500 ml, Plastic bottle
CAS Number:  
Price
$319.87
Log in pour la disponibilité
Each (500ml) Case of 12 (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Spécifications
Spécifications
  • Type de conditionnement:
    Plastic bottle
  • Taille:
    500 ml