Certipur® TOC standard solution acc. to EN 1484-H3/DIN 38409-H3 as potassium hydrogen phthalate in water, stabilized
CA1.09017.0100
:
- Concentration:1000 mg/L
- Pack type:Glass Bottle
- Description:TOC standard
- Size:100 ml
- Cat. no.:CA1.09017.0100
- Supplier No.:1.09017.0100
- Diluted standard solutions that are ready-to-use for accurate measurements
- Traceable to SRM from NIST (SRM 84L)
- Full certificates of analysis with comprehensive quality documentation
- Information on concentration and expanded measurement uncertainty
- Save time and minimize errors
: Store at +15 to +25 °C.