Di-Sodium Hydrogen Phosphate Solution (Buffer Stock Solution), Certipur® | Avantor
Di-Sodium Hydrogen Phosphate Solution (Buffer Stock Solution), Certipur®
di-Sodium hydrogen phosphate solution, 1/15 mol/l, Phosphate, 20 °C, Plastic bottle, 1 L, 9.2
Catalog # CA1.06587.1000
Supplier:   MilliporeSigma
Fournisseur #:  1.06587.1000
Di-Sodium Hydrogen Phosphate Solution (Buffer Stock Solution), Certipur®
Catalog # CA1.06587.1000
Supplier:  MilliporeSigma
CAS Number:  
Di-Sodium Hydrogen Phosphate Solution (Buffer Stock Solution), Certipur®
Di-Sodium Hydrogen Phosphate Solution (Buffer Stock Solution), Certipur®
di-Sodium hydrogen phosphate solution, 1/15 mol/l, Phosphate, 20 °C, Plastic bottle, 1 L, 9.2
CAS Number:  
Price
$129.48
Log in pour la disponibilité
Each (1l) Case of 6 (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Buffer stock solution 1/15 mol/l
Spécifications
Spécifications
  • Type de conditionnement:
    Plastic bottle
  • Désignation:
    di-Sodium hydrogen phosphate solution, 1/15 mol/l
  • Taille:
    1 L
  • pH Buffer Type:
    Phosphate
  • Reference Temperature:
    20 °C
  • pH:
    9.2