Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck | Avantor
Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck
Supplier:   MilliporeSigma
Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck
Supplier:  MilliporeSigma
Certificates
Aluminium Oxide 60 G Neutral (type E) for Thin-Layer Chromatography, Merck
1 Options Available Below
Product Details & Documents
Standardized sorbents ensure reliable results every time
EMD Millipore's broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µ: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).
Product Family Options
Informations sur le produit
  • Taille
    Type de conditionnement
    Availability
    Price
  • 2.5 kg
    Plastic Bottle
    Discontinued
    Supplier Number: 1.01090.2500
  • 2.5 kg
    Plastic Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (2,500g)
    Each (2,500g) - 3 814,11 $
    3 814,11 $
    Log in pour la disponibilité
    Case of 4 (2,500,g)
    Case of 4 (2,500,g) - 15 256,44 $
    15 256,44 $
    Each (2,500g)
       3 814,11 $
    Case of 4 (2,500,g)
       15 256,44 $
    Supplier Number: 1.01090.2500