24/40 Dean-Stark Moisture Test Receiver, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
With return line to allow excess solvent to return to boiling flask
Suitable for ASTM D 95 and E 123. Joints are a standard taper 24/40.
Frequently Bought Together