Optimize Back Pressure Regulators
Supplier:   Optimize
Optimize Back Pressure Regulators
Supplier:  Optimize
Certificates
Optimize Back Pressure Regulators
Optimize Back Pressure Regulators
5 Options Available Below
Product Details & Documents
Optimize flow-through back pressure regulators utilize an active mechanism to maintain steady back pressure across a range of mobile phase viscosities and flow rates.
  • Flow-through design
  • Utilizes an active mechanism to maintain steady back pressure across a range of conditions
  • Prevent solvent decompression and bubble formation in the flow cell
  • Reduces baseline noise and drift
  • 100 µl internal volume
Placing one downstream from your detector will prevent solvent decompression and bubble formation in the flow cell, reducing baseline noise and drift. Each pre-set Optimize stainless steel Back Pressure Regulator offers a 100 µl internal volume and includes fittings for immediate connection out of the box.
 : Includes OPTI-LOK® LP Fittings.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • 10 psi back pressure regulator
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $888.46
    $888.46
    Each
       $888.46
    Supplier Number: 10-06-00120
  • 30 psi back pressure regulator
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $885.89
    $885.89
    Each
       $885.89
    Supplier Number: 10-06-00128
  • 60 psi back pressure regulator
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,010.05
    $1,010.05
    Each
       $1,010.05
    Supplier Number: 10-06-00129
  • 100 psi back pressure regulator
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $898.89
    $898.89
    Each
       $898.89
    Supplier Number: 10-06-00130
  • 150 psi back pressure regulator
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $885.89
    $885.89
    Each
       $885.89
    Supplier Number: 10-06-00132