Dual Position Closure Caps, Globe Scientific
Fournisseur: Globe Scientific
Certificats
À propos de cet article
This dual position cap is produced from low density polyethylene (LDPE). It is ribbed for easy handling.
The cap is specially designed to provide aerobic or anaerobic storage of culture samples. When snapped into the tube, the cap provides a leak-resistant seal.
Note: Leave the cap in the loose position for aerobic work and in the sealed position for anaerobic cultures.
Frequently Bought Together