Wall Mounted Drying Rack, Globe Scientific | Avantor
Wall Mounted Drying Rack, Globe Scientific
Wall Mounted Drying Rack, 19.6", 19.6"
Catalog # 89498-072
Supplier:   Globe Scientific
Fournisseur #:  600213
Wall Mounted Drying Rack, Globe Scientific
Catalog # 89498-072
Supplier:  Globe Scientific
Wall Mounted Drying Rack, Globe Scientific
Wall Mounted Drying Rack, Globe Scientific
Wall Mounted Drying Rack, 19.6", 19.6"
Price
$982.80
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
This 72-place wall mountable laboratory drying rack is ideal for draining and drying all types of Labware
The back plate of the support is made from a single mold of 4 mm rigid high-impact polystyrene (HIPS). The peg sockets feature closed ends, eliminating leakage and consequent biohazards. The seamless drip channel collects waste which drains via a supplied drain tube.
The 72 (push in) pegs can accommodate items with a 15 mm+ neck bore. They are easily removable for cleaning or to accommodate oddly shaped items. Also included are 11 small pegs to accommodate small diameter items with a neck bore of 6mm+ (such as test tubes). This durable rack will not rust or stain.
The rack is supplied with a mounting kit to facilitate convenient hanging.
Spécifications
  • Largeur:
    19.6"
  • Désignation:
    Wall Mounted Drying Rack
  • Hauteur:
    19.6"
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Extra Pegs for Drying Rack, Small Diameter, 95 x 6 mm neck bore of 6 mm
    Extra Pegs for Drying Rack, Small Diameter, 95 x 6 mm neck bore of 6 mm
    10755-178
    Log in pour la disponibilité
    Pack of 11 - $66.85