Large Scale Evaporator Receiving Flask, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Receiving flasks are designed to fit all large-scale rotary evaporators
Now available in coated, plain, non-coated, amberized, and jacketed versions. Side necks include 89214-958 SVL-22 threaded black vent caps. Top and bottom socket joints are DN25, and bottom joint includes 89061-126 CAPFE (PTFE-encapsulated silicone rubber) o-ring, size –217.
Frequently Bought Together