Polystyrene and Polypropylene Test Tubes, 17×100 mm, Globe Scientific
Supplier: Globe Scientific
Certificates
About this item
These tubes are molded either from premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis and solvents.
Caps are made of polyethylene and are not autoclavable.
Frequently Bought Together