ContactUsLinkComponent
Jacketed Evaporator Receiving Flask, Ace Glass Incorporated
Catalog # 89215-046
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Jacketed Evaporator Receiving Flask, Ace Glass Incorporated
Catalog # 89215-046
Supplier:  Ace Glass
Supplier Number:  6727-10
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Capacity:
    8 L
  • Description:
    Receiving Flask, Jacketed, 8 L
  • Cat. no.:
    89215-046
  • Fits Models:
    All R220
  • Supplier No.:
    6727-10
Specifications
About this item
New design, jacketed receiving flask
Fits all rotary evaporators. Outer jacket has 28/15 o-ring ball joints to seal to chiller hoses. Has an SVL thread vent cap on side joint. Bottom DN25 ball joint has CAPFE o-ring. Jacket inlet and outlet ball joints have FETFE o-rings