CritiCore ESD Stripe Cleanroom Hoods with Open Face Below Lip
Supplier: VWR International
Certificates
About this item
These reusable cleanroom hoods are made with a high-density polyester calendered fabric that resists fluids and bacterial penetration.
- Hoods are a one-size-fits-all pullover
- Tunneled elastic back plus horizontal and vertical adjustment snaps
- Split-drape design with a front panel longer than the back
- All seams are laser-cut or pre-serged
- Thread is silicon-free
Designed for cleanroom applications up to and including Class 10 (ISO 4), hoods also include a 1% carbon stripe for electrostatic dissipation. Suitable applications include pharmaceutical, bio-tech, and medical device manufacturing.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Specifications
- Catalog No:
- 89495-810
- 89495-808
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Risk area:
- Anti-static, ESD
- Anti-static, ESD
- Gender:
- Unisex
- Unisex
- Closure type:
- Back fastening
- Back fastening
- Fabric:
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- Weight:
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.