CritiCore Cleanroom Frocks with ESD Stripe
Fournisseur: VWR International
Certificats
À propos de cet article
Reusable zippered frocks are designed for controlled environments and less critical cleanroom applications
- Garments feature a mandarin collar with two snaps at top, a wide-pull zipper, and single snap below at bottom
- Assembled with double-needle felled seams for added durability and particle migration prevention
- All seams are laser-cut or pre-serged, and thread is silicon-free
- Available in white and navy
Made with lightweight yet durable high-density ESD striped fabric, they resist fluid and bacterial penetration. A 1% carbon stripe provides electrostatic dissipation, and knit cuffs include 5% carbon Belltron fiber. Applications include pharmaceutical, bio-tech, and medical device manufacturing.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Spécifications
- Catalog No:
- 89495-736
- 89495-758
- 89495-738
- 89495-750
- 89495-752
- 89495-754
- 89495-756
- 89495-764
- 89495-742
- 89495-766
- 89495-744
- 89495-746
- 89495-768
- 89495-748
- 89495-770
- 89495-760
- 89495-762
- 89495-740
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- ESD Stripe (99% polyester, 1% carbon)
- Weight:
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.