WaterPro® RO Systems, Labconco
À propos de cet article
WaterPro RO Stations feature a prefilter, carbon filter and RO membrane that combine to deliver high-quality reverse osmosis (RO) purified water
WaterPro RO may be connected to laboratory glassware washers, such as SteamScrubber® or FlaskScrubber® Vantage, to provide purified rinse water. Features include a 17L tank, Type 304 stainless steel front panel and control panel.
RO purified water is ideal for laboratory applications such as reagent preparation and glassware rinsing. Water may be dispensed manually from a valve or from an optional dispensing gun (sold separately).
: Dispensing gun not included. Prefilter/Carbon Filter Kit 9067201 and Reverse Osmosis Membrane 9078700 are required, but sold separately.
6 Options Available Below
Product Details & Documents
WaterPro RO Stations feature a prefilter, carbon filter and RO membrane that combine to deliver high-quality reverse osmosis (RO) purified water
WaterPro RO may be connected to laboratory glassware washers, such as SteamScrubber® or FlaskScrubber® Vantage, to provide purified rinse water. Features include a 17L tank, Type 304 stainless steel front panel and control panel.
RO purified water is ideal for laboratory applications such as reagent preparation and glassware rinsing. Water may be dispensed manually from a valve or from an optional dispensing gun (sold separately).
: Dispensing gun not included. Prefilter/Carbon Filter Kit 9067201 and Reverse Osmosis Membrane 9078700 are required, but sold separately.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 9075020
- Supplier Number: 9075030
- Supplier Number: 9075070
- Supplier Number: 9075080
- Supplier Number: 9075060
- Supplier Number: 9075040