ContactUsLinkComponent
SP Bel-Art Screw Style Tubing Clamp, Bel-Art Products, a part of SP
SP Bel-Art Screw Style Tubing Clamp, Bel-Art Products, a part of SP
Catalog # 70680-322
undefined
SP Bel-Art Screw Style Tubing Clamp, Bel-Art Products, a part of SP
Catalog # 70680-322
Supplier Number:  F18212-0000
Specifications
  • Cat. no.:
    70680-322
  • Fits Tubing O.D.:
    max. 14 mm (¹/₂")
  • Supplier No.:
    182120000
Specifications
About this item
Offers on/off and fine control so accurate that it replaces stopcocks in many applications.
  • Molded acetal plastic construction
  • Very fine, accurate control
  • Autoclavable
  • Resistant to acids, alkalis, and organic solvents
Can be installed without disconnecting tubing or interrupting operation. Molded of Acetal plastic, the clamp is autoclavable and resistant to acids, alkalis and organic solvents. Accepts flexible tubing up to 14 mm (½") O.D.