Sample Cups for Abbott Analyzer, Globe Scientific | Avantor
Sample Cups for Abbott Analyzer, Globe Scientific
Supplier:   Globe Scientific
Sample Cups for Abbott Analyzer, Globe Scientific
Supplier:  Globe Scientific
Certificates
Sample Cups for Abbott Analyzer, Globe Scientific
Sample Cups for Abbott Analyzer, Globe Scientific
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These sample cups and sample tubes are designed for use on Abbott® Axsym analyzers.
Product Family Options
Informations sur le produit
  • Désignation
    Hauteur
    Ø ext.
    Availability
    Price
  • Sample Tube
    75 mm
    16 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $269.57
    $269.57
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $269.57
    Supplier Number: 5543
  • Sample Cup
    53 mm
    17.5 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $146.15
    $146.15
    Prix promotionnel pour les nouveaux clients
    Pack of 1,000
       $146.15
    Supplier Number: 5542