Nesting Sample Cups, Globe Scientific
À propos de cet article
Designed for use in conjunction with other test tubes or original blood collection tubes when smaller sampling is necessary.
- Polystyrene (PS)
- Excellent clarity
- Temperate Range: –10 to 50 °C
- Non-sterile
To use, simply transfer the sample from the original collection tube into the nesting cup. Then, place the nesting cup inside the original collection tube. The nesting cup "rides" along with the originally labeled/barcoded tube in the analyzer. This procedure saves time by eliminating the need to re-label the smaller sample.
2 Options Available Below
Product Details & Documents
Designed for use in conjunction with other test tubes or original blood collection tubes when smaller sampling is necessary.
- Polystyrene (PS)
- Excellent clarity
- Temperate Range: –10 to 50 °C
- Non-sterile
To use, simply transfer the sample from the original collection tube into the nesting cup. Then, place the nesting cup inside the original collection tube. The nesting cup "rides" along with the originally labeled/barcoded tube in the analyzer. This procedure saves time by eliminating the need to re-label the smaller sample.
Product Family Options
Informations sur le produit
- CapacitéDésignationHauteurFor Use WithMatièreØAvailabilityPrice
- 1 mlNesting Sample Cup30 mm12mm and 13mm TubesPolystyrene13 mmLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $102.61$102.61Prix promotionnel pour les nouveaux clientsPack of 1,000$102.61Supplier Number: 5504
- 2 mlNesting Sample Cup30 mm16 mm TubesPolystyrene16 mmLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $167.08$167.08Prix promotionnel pour les nouveaux clientsPack of 1,000$167.08Supplier Number: 5505