VWR® Slider Top Reclosable Bags
Fournisseur: VWR International
Certificats
À propos de cet article
These three zip top slider bags are great for applications where quick, positive closure and ease of use are important.
- Zip top design
- Quick, positive closure
- Easy to use
- Made of 100% virgin low density polyethylene (LDPE) resin
Ideal for use with gloved hands. The slider helps hold the bag open for one handed loading.
Certifications: Meet FDA/USDA specifications.
Frequently Bought Together