Universal Snap Cap, Globe Scientific
Universal Snap Cap, Globe Scientific
Supplier:   Globe Scientific
Universal Snap Cap, Globe Scientific
Supplier:  Globe Scientific
Certificates
Universal Snap Cap, Globe Scientific
Universal Snap Cap, Globe Scientific
7 Options Available Below
Product Details & Documents
Guard samples against cross-contamination and laboratory work areas against infection and spillage with these universal snap caps
  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Add the convenience of 1 cap to fit multiple size tubes
These polyethylene universal caps are designed for easy-on and easy-off use. Cap removal is simple and quick. Universal caps are ideal for recapping most 12 mm, 13 mm, and 16 mm evacuated glass blood collection tubes. Flexible closures protect from aerosols of highly infectious microorganisms.
Precision molded from low-density polyethylene and designed to fit firmly on the tube. Closures are very sturdy under most rigorous procedures, including centrifugation.
Ordering information: Snap caps are supplied in a pack of 1000.
Product Family Options
Product Information
  • Désignation
    Matière
    Color
    Availability
    Price
  • Universal Snap Cap
    Polyethylene
    Black
    Log in for availability
    Log in for availability
    Bag of 1,000
    Bag of 1,000 - $90.40
    $90.40
    New Customer Promotional Price
    Bag of 1,000
       $90.40
    Supplier Number: 118115K
  • Universal Snap Cap
    Polyethylene
    Blue
    Log in for availability
    Log in for availability
    Bag of 1,000
    Bag of 1,000 - $89.98
    $89.98
    New Customer Promotional Price
    Bag of 1,000
       $89.98
    Supplier Number: 118115B
  • Universal Snap Cap
    Polyethylene
    Green
    Log in for availability
    Log in for availability
    Bag of 1,000
    Bag of 1,000 - $90.40
    $90.40
    New Customer Promotional Price
    Bag of 1,000
       $90.40
    Supplier Number: 118115G
  • Universal Snap Cap
    Polyethylene
    Lavender
    Log in for availability
    Log in for availability
    Bag of 1,000
    Bag of 1,000 - $87.61
    $87.61
    New Customer Promotional Price
    Bag of 1,000
       $87.61
    Supplier Number: 118115L
  • Universal Snap Cap
    Polyethylene
    Natural
    Log in for availability
    Log in for availability
    Bag of 1,000
    Bag of 1,000 - $85.14
    $85.14
    New Customer Promotional Price
    Bag of 1,000
       $85.14
    Supplier Number: 118115C
  • Universal Snap Cap
    Polyethylene
    Red
    Log in for availability
    Log in for availability
    Bag of 1,000
    Bag of 1,000 - $90.40
    $90.40
    New Customer Promotional Price
    Bag of 1,000
       $90.40
    Supplier Number: 118115R
  • Universal Snap Cap
    Polyethylene
    Yellow
    Log in for availability
    Log in for availability
    Bag of 1,000
    Bag of 1,000 - $88.45
    $88.45
    New Customer Promotional Price
    Bag of 1,000
       $88.45
    Supplier Number: 118115Y