Unmethylated Lambda DNA, New England Biolabs | Avantor
Unmethylated Lambda DNA, New England Biolabs
Unmethylated Lambda DNA, New England Biolabs
Certificates
Unmethylated Lambda DNA, New England Biolabs
Unmethylated Lambda DNA, New England Biolabs
2 Options Available Below
Product Details & Documents
Duplex DNA is isolated from bacteriophage lambda (cI857ind 1 Sam 7)
Therefore, those deoxyadenosine positions which are methylated in a wild type E. coli host are not methylated in the dam- host. This DNA gives complete digests with restriction endonucleases that are sensitive to dam methylation. Lambda DNA (N6-methyladenine-free) is 48,502 base pairs in length. The phage is isolated from a heat-inducible E. coli (dam-) lambda lysogen. The DNA is isolated from purified phage by phenol extraction and dialyzed against 10 mM Tris-HCl (pH 8.0) and 1 mM EDTA.
The bacteriophage is grown in an E. coli host which is deficient in adenine methylase (dam-).
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Unmethylated Lambda DNA
    1,250 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $492.00
    $492.00
    Each
       $492.00
    La quantité minimale de commande est de 1
    Supplier Number: N3013L
  • Unmethylated Lambda DNA
    250 µg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $125.99
    $125.99
    Each
       $125.99
    La quantité minimale de commande est de 1
    Supplier Number: N3013S