À propos de cet article
The HiScribe T7 High Yield RNA Synthesis Kit is an extremely flexible system for in vitro transcription of RNA using T7 RNA Polymerase.
The kit allows for synthesis many kinds of RNA including internally labeled and co-transcriptionally capped transcripts. RNA synthesized from the kit is suitable for many applications including RNA structure and function studies, ribozyme biochemistry, probes for RNase protection assays and hybridization based blots, anti-sense RNA and RNAi experiments, microarray analysis, microinjection, and in vitro translation and RNA vaccines. The kit contains sufficient reagents for 50 reactions of 20 μl each. Each standard reaction yields up to 180 μg of RNA from 1 μg control template. Each kit can yield up to 9 mg RNA. For 32P labeling, the kit contains enough reagents for 100 reactions of 20 μl each.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.