Supercoiled DNA Ladder , New England Biolabs
Catalog # CA101449-342
Supplier: New England Biolabs (NEB)
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Supercoiled DNA Ladder
- Size:100 gel lanes
- Cat. no.:CA101449-342
Specifications
About this item
The Supercoiled DNA ladder contains 9 proprietary supercoiled plasmids, ranging in size from 2 to 10 kb, that are suitable for use as supercoiled molecular weight standards for agarose electrophoresis
The 5 kb plasmid has an increased intensity to serve as a reference band. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.