Standard Taq Reaction Buffer Pack, New England Biolabs
Catalog # CA101227-506
Supplier: New England Biolabs (NEB)
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Standard Taq Reaction Buffer Pack
- Cat. no.:CA101227-506
- Volume:6.0 ml
Specifications
About this item
New England Biolabs supplies a unique 10X reaction buffer for use with Taq DNA Polymerase in reaction systems which require a detergent free, low ionic strength buffer.