SP6 RNA Polymerase, New England Biolabs
Catalog # CA101228-202
Supplier: New England Biolabs (NEB)
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:qPCR probe
- Description:SP6 RNA Polymerase
- Size:2,000 units
- Cat. no.:CA101228-202
Specifications
About this item
Bacteriophage SP6 RNA Polymerase is a DNA-dependent RNA polymerase that is highly specific for the SP6 phage promoter
The 98.5 KD polymerase catalyzes in vitro RNA synthesis from a cloned DNA template under the SP6 promoter. RNA synthesized using the SP6 RNA Polymerase is suitable for many applications in research and biotechnology.