ContactUsLinkComponent
SNAP-BIOTIN®, New England Biolabs
Catalog # CA101447-668
CAS Number:  
undefined
SNAP-BIOTIN®, New England Biolabs
Catalog # CA101447-668
Supplier Number:  S9110S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    SNAP-Biotin
  • Size:
    50 nmol
  • Cat. no.:
    CA101447-668
Specifications
About this item
SNAP-Biotin®i s a cell-permeable substrate (BG-Biotin) based on biotin with an amidocaproyl linker
It is suitable for applications such as biotinylation of SNAP-tag® fusion proteins in living cells for detection with streptavidin fluorophore conjugates or labeling in solution for analysis by SDS-PAGE/Western blot or for capture with streptavidin for binding and interaction studies. This package contains 50 nmol of SNAP-Biotin substrate, sufficient to make 10 ml of a 5 μM solution for the labeling of SNAP-tag fusion proteins in cells.