RNase HII
Fournisseur: New England Biolabs (NEB)
Certificats
À propos de cet article
Ribonuclease HII (RNase HII) is an endoribonuclease that preferentially nicks 5´ to a ribonucleotide within the context of a DNA duplex
The enzyme leaves 5´ phosphate and 3´ hydroxyl ends. RNase HII will also nick at multiple sites along the RNA portion of an Okazaki fragment.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Enzyme name:RNase HII