ContactUsLinkComponent
PUREXPRESS® delta Ribosome Kit, New England Biolabs
Catalog # CA101640-812
CAS Number:  
undefined
PUREXPRESS® delta Ribosome Kit, New England Biolabs
Catalog # CA101640-812
Supplier Number:  E3313S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    PURExpress delta Ribosome Kit
  • Size:
    10 Reactions
  • Cat. no.:
    CA101640-812
Specifications
About this item
A rapid method for gene expression analysis, PURExpress® is a novel cell-free transcription/translation system reconstituted from the purified components necessary for E
The relative nuclease-free and protease-free nature of the PURExpress platform preserves the integrity of DNA and RNA templates/ complexes and results in proteins that are free of modification and degradation. Transcription and translation are carried out in a one-step reaction, and require the mixing of only two tubes. With results available in a few hours, PURExpress saves valuable laboratory time and is ideal for high throughput technologies. The ribosomes are removed from this kit, allowing users to add their own modified ribosomes for studies on protein translation. Control ribosomes (sufficient for 2 reactions) are provided in a separate tube.
coli translation.