To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:PURExpress (aa, tRNA) Kit
- Size:10 Reactions
- Cat. no.:CA101641-562
A rapid method for gene expression analysis, PURExpress® is a novel cell-free transcription/translation system reconstituted from the purified components necessary for E
The relative nuclease-free and protease-free nature of the PURExpress platform preserves the integrity of DNA and RNA templates/ complexes and results in proteins that are free of modification and degradation. Transcription and translation are carried out in a one-step reaction, and require the mixing of only two tubes. With results available in a few hours, PURExpress saves valuable laboratory time and is ideal for high throughput technologies. The tRNA and amino acids are supplied separately in this kit, allowing users to run a protein synthesis reaction by adding modified amino acids and tRNA mixtures to the reaction.
coli translation.